Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLC29A4 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5637R-CY5.5
BOSSBS-5637R-CY5.5EA 1350 EUR InStock
BOSSBS-5637R-CY5.5
Anti-SLC29A4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SLC29A4 (Solute carrier family 29 (nucleoside transporters), member 4) is involved in the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It transports serotonin, dopamine and the neurotoxin 1-methyl-4-phenylpyridinium. It may also contribute to the regulation of extracellular adenosine concentrations.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC29A4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge