Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CABLES2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5744R-CY5.5
BOSSBS-5744R-CY5.5EA 900 EUR InStock
BOSSBS-5744R-CY5.5
Anti-CABLES2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
CABLES2 is a proapoptotic factor involved in both p53-mediated and p53-independent apoptotic pathways. ik3-1/CABLES1, a closely related molecule, has been identified to be a candidate tumor suppressor for colon and head/neck cancers.The exact function of this protein is unknown, but it is probably involved in G1-S cell cycle transition. CABLES2 binds to CDK3, CDK5 and ABL1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CABLES2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge