Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PROM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5814R-A555
BOSSBS-5814R-A555EA 915 EUR InStock
BOSSBS-5814R-A555
Anti-PROM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Prominin 2 is a 112 kDa glycoporotein structurally related to Prominin 1 (CD133) although amino acid similarity is not more than 30%, but their genomic organization is strikingly similar. Like Prominin 1, the prominin 2 exhibit similar membrane topology with 5 trans-membrane domains and two large glycosylated extracellular domains. Similar to Prominin1 localization, the Prominin 2 is also associated with membrane protrusions of the epithelial cells from adult kidney, and all along the digestive track and other epithelial tissues.Prominin 2 expression is down-regulated in aggressive prostate cancer cell lines and transient transfection of PROML2 expression vectors has been shown to induce apoptosis in cultured prostate cancer cells, suggesting a tumor suppressive role for Prominin 2. Prominin 2 expression is likely to be involved in growth suppression in the prostate, and down-regulation of Prominin 2 may disrupt normal prostatic homeostasis and lead to uncontrolled prostatic growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PROM2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge