Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADAMTS8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5859R-A488
BOSSBS-5859R-A488EA 893 EUR InStock
BOSSBS-5859R-A488
Anti-ADAMTS8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ADAMTS proteases are secreted enzymes containing a prometalloprotease domain of the reprolysin type. The ADAMTS proteases function in processing of procollagens and von Willebrand factor as well as catabolism of aggrecan, versican and brevican. They have been demonstrated to have important roles in connective tissue organization, coagulation, inflammation, arthritis, angiogenesis and cell migration.A member of the metalloproteinase family containing disintegrin like domains (ADAMs), the function of ADAMTS8 is still poorly understood. ADAMTS8 contains the canonical HExxHxxxxxH zinc metalloproteinase motif, and has been shown to be proteolytically active on a range of substrates. ADAMTS8 is inhibited by the endogenous MMP inhibitors, TIMP1, 2, 3 and 4, but most efficiently by TIMP3. In addition to the metalloprotease domain, ADAMTS8 has a propeptide domain, a Prohormone Convertase (PC, furin) cleavage site, a cysteine rich domain and thrombospondin 1 like domains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTS8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge