Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADAMTSL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5862R-A488
BOSSBS-5862R-A488EA 893 EUR InStock
BOSSBS-5862R-A488
Anti-ADAMTSL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ADAMTS (A Disintegrin And Metalloproteinase Domain with Thrombospondin type 1 Modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer. ADAMTS protein family members contain an amino-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a carboxy-terminus that contains a varying number of Thrombospondin type 1 (TSP-1) motifs. ADAMTS-L2 (ADAMTS-like protein 2) is a 951 amino acid secreted protein that is highly expressed in lung, kidney and liver. Mutations in the gene encoding ADAMTS are the cause of geleophysic dysplasia, an autosomal recessive disorder characterized by cardiac vavular anomalies, short stature, thick skin and brachydactyly. In individuals affected with geleophysic dysplasia, there is a significant increase in total active TGF-beta 1 and nuclear locations of p-SAMD2 in fibroblasts. Interestingly, ADAMTS-L2 interacts with LTBP-1, a glycoprotein that is part of the platelet-derived TGF-beta 1 complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTSL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge