Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-WAVE1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5895R-CY3
BOSSBS-5895R-CY3EA 900 EUR InStock
BOSSBS-5895R-CY3
Anti-WAVE1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
WASP (for Wiskott-Aldrich syndrome protein) and N-WASP are downstream effectors of Cdc42 that are implicated in Actin polymerization and cytoskeletal organization. The WASP family also includes VASP (vasodilator-stimulated phosphoprotein) and Mena (for mammalian enabled protein), which accumulate at focal adhesions and are also involved in the regulation of the Actin cytoskeleton. The WAVE proteins are related to the WASP family proteins and are likewise involved in mediating Actin reorganization downstream of the Rho family of small GTPases. The protein homologs WAVE1 and WAVE2 regulate membrane ruffling by inducing the formation of Actin filament clusters in response to GTP binding and by activating Rac. They mediate Actin polymerization by cooperating with the Arp2/3 complex, thereby promoting the formation of Actin filaments. WAVE1, which is also designated SCAR (suppressor of cAR), is expressed primarily in the brain, while WAVE2 is widely expressed, with the expression highest in peripheral blood leukocytes. WAVE3 forms a multiprotein complex that links receptor kinases with Actin and plays a role in the transduction of signals involving changes in cell shape, function or motility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WAVE1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge