Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ANGPTL3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5918R-A350
BOSSBS-5918R-A350EA 893 EUR InStock
BOSSBS-5918R-A350
Anti-ANGPTL3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Angiopoietin-like protein 3 (Angptl3) functions as a potent lipoprotein lipase inhibitor and is an important component of plasma triglyceride homeostasis. Angptl3 also plays a role in adipose formation and angiogenesis through its interaction with integrin ?v)beta(3). It is secreted by the liver and is functionally defined by the C-terminal fibrinogen (FBN)-like domain and an N-terminal coiled-coil domain. Angptl3 regulates circulating triglyceride levels during different nutritional states thereby mediating the feeding/fasting cycle. A deficiency of Angptl3 results in abnormally low lipid levels, and a repression of the protein may be protective against atherosclerosis. Angptl3 may also play an important role in hyperlipidemia in diabetes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge