Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-POMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5952R-A555
BOSSBS-5952R-A555EA 893 EUR InStock
BOSSBS-5952R-A555
Anti-POMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
POMT1 (Protein O mannosyl transferase 1) is a multipass membrane protein that is found in the endoplasmic reticulum. POMT1 catalyses the transfer of mannosyl residues to the hydroxyl groups of serine or threonine residues. Enzymatic activity is dependent on co expression of POMT1 with POMT2. Defects in the POMT1 gene are associated with Walker-Warburg syndrome (WWS), a congential muscular dystrophy that is associated with mental retardation and is usually lethal within the first few months of life. Other defects in the POMT1 gene result in limb girdle muscular dystrophy type 2K (LGMD2K), which is associated with mild mental retardation. Studies in Drosophila suggest that mutation of POMT1 alters the efficacy of synaptic transmission and a change in subunit composition of post synaptic glutamate receptors at the neuromuscular junction. Missense mutations in POMT1 have been associated with glioneuronal and glial brain tumours.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POMT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge