Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDCA7L Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5963R-CY5.5
BOSSBS-5963R-CY5.5EA 900 EUR InStock
BOSSBS-5963R-CY5.5
Anti-CDCA7L Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
CDCA7L is a member of the CDCA (cell division cycle associated) protein family. Members of this family have expression patterns associated with other known cell cycle genes such as cyclins and CDC genes. CDCA7L plays a role in transcriptional regulation as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to the promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It is involved in apoptotic signaling pathways and may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA7L
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge