Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SMYD3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5974R-CY3
BOSSBS-5974R-CY3EA 900 EUR InStock
BOSSBS-5974R-CY3
Anti-SMYD3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
SMYD3 is a gene that is over-expressed in the majority of colorectal carcinomas and hepatocellular carcinomas. SMYD3 forms a complex with RNA polymerase II through an interaction with the RNA helicase HELZ and transactivates a set of genes that included oncogenes, homeobox genes and genes associated with cell-cycle regulation. SMYD3 binds to a motif, 5'-CCCTCC-3', present in the promoter region of downstream genes such as Nkx2.8. The SET domain of SMYD3 shows histone H3-lysine 4 (H3-K4)-specific methyltransferase activity, which is enhanced in the presence of the heat-shock protein HSP90A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMYD3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge