Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FSTL1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6050R-CY5.5
BOSSBS-6050R-CY5.5EA 900 EUR InStock
BOSSBS-6050R-CY5.5
Anti-FSTL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
TSC-36 is a secreted extracellular glycoprotein. The amino acid sequence of TSC-36 is similar to follistatin, an inhibitor of activin, as it contains a follistatin module. TSC-36 is a heparin-binding protein suggested to have a role in the negative regulation of cellular growth, as its expression is induced in response to TGF-b1. In addition, TSC-36 is not found in small cell lung cancer (SCLC) cells, a highly aggressive neoplasm, but is detected in some non-small cell lung cancer (NSCLC) cells, a moderately aggressive neoplasm.. May modulate the action of some growth factors on cell proliferation and differentiation. Binds heparin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge