Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-XAB2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6080R-CY5.5
BOSSBS-6080R-CY5.5EA 900 EUR InStock
BOSSBS-6080R-CY5.5
Anti-XAB2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
XAB2 protein (XPA binding protein 2) was first identified through its interaction with XPA, a key factor in nucleotide excision repair pathways. XAB2 therefore appears to be involved in transcription coupled DNA repair and transcription. Immunoprecipitation experiments have also demonstrated that a fraction of XAB2 interacts with the transcription coupled repair specific proteins CSA and CSB as well as with RNA polymerase II. Microinjection experiments with XAB2 resulted in the inhibition of both transcription coupled repair and transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: XAB2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge