Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RIN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6094R-HRP
BOSSBS-6094R-HRPEA 900 EUR InStock
BOSSBS-6094R-HRP
Anti-RIN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
RIN1 is a Ras effector protein which can affect Ras signaling in a variety of ways. It enhances signaling from ABL1 and ABL2 whcih are involved in regulation of cytoskeletal remodeling. It may function as a guanine nucleotide exchange factor in activation of Rab5a. Rin1 also competes with RAF1 for binding of the activated form of Ras. It may play a role inhibiting the modulation of neuronal plasticity in formation of aversive memory, and has been implicated as a tumor supressor gene in breast cancer.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RIN1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge