Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RND3 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6095R
BOSSBS-6095REA 725 EUR InStock
BOSSBS-6095R
Anti-RND3 Rabbit Polyclonal Antibody
Antikörper
The Rho family of proteins mediates cytoskeleton organization and cell motility responses. This family contains twenty different members divided into five groups, the Rho-like, Rac-like, Cdc42-like, Rnd and RhoBTB subfamilies. Rho proteins have GTPase activity and bind GTP in their active form and GDP in their inactive form. In their active form they interact with downstream proteins. However, the Rnd subfamily of proteins containing the Rnd1, Rnd2 and RhoE(Rnd3), binds GTP but not GDP and has no GTPase activity. While Rho proteins are required for stress fibre formation in cultured fibroblasts, epithelial and endothelial cells, overexpression of Rnd3 or Rnd1 in the same cells may cause an opposite effect (i.e. decrease in stress fibre). Thus, the Rnd3 protein may act as an antagonist for the Rho proteins either by directly binding to Rho or to other downstream proteins.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge