Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RBBP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6130R-HRP
BOSSBS-6130R-HRPEA 900 EUR InStock
BOSSBS-6130R-HRP
Anti-RBBP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
RbAp48 (Retinoblastoma-binding protein p48 or Rb-associated protein p48)is a WD repeat protein that is a core histone binding subunit common to several complexes involved in chromatin assembly, chromatin remodeling, and histone deacetylation. These complexes include Chromatin Assembly Factor 1 (CAF1), which is required for chromatin assembly following DNA replication and repair, the core histone deacetylase (HDAC) complex, which promotes histone deacetylation and subsequent transcriptional repression, the nucleosome remodeling and histone deacetylation complex NuRD, the nucleosome remodeling factor (NURF) complex, and the PRC2 complex, which promotes repression of homeotic genes during development. RpAp48 also interacts with the retinoblastoma protein, and with SPEN/MINT and BRCA1. It is also a component of the DREAM complex, which represses cell cycle-dependent genes in quiescent cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RBBP4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge