Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VAV2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6160R-CY7
BOSSBS-6160R-CY7EA 900 EUR InStock
BOSSBS-6160R-CY7
Anti-VAV2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
VAV2 is a ubiquitously expressed structural homolog of the VAV protooncogene that is expressed preferentially in hematopoetic cells. Both proteins are comprised of a Dbl homology (DH) domain with guanosine nucleotide exchange (GEF) activity exclusively directed towards Rho/Rac GTPases, a pleckstrin homology (PH) domain, a calponin-homology (CH) region, an acidic domain (AD) a zinc finger butterfly motif, two SH3 regions and one SH2 domain. GEF activity of RhoA family G proteins is induced by tyrosine phosphorylation in wild type VAV2, and is constitutively activated in N terminus deleted oncogene forms. Constitutive expression of a VAV2 oncoprotein may result in morphological alterations including highly enlarged cells in which karyokinesis and cytokinesis frequently are uncoupled.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VAV2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge