Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Rab3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6180R-CY5.5
BOSSBS-6180R-CY5.5EA 900 EUR InStock
BOSSBS-6180R-CY5.5
Anti-Rab3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has four known isoforms: 3A, 3B, 3C and 3D. These proteins are expressed in the highest concentration in the brain and endocrine tissues and are involved in regulated exocytosis of hormones and neurotransmitters. Specifically Rab3's are thought to play a role in presynaptic vesicle trafficking and priming.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Rab3A+Rab3B+Rab3C+Rab3D
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge