Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TMPRSS4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6286R-A350
BOSSBS-6286R-A350EA 915 EUR InStock
BOSSBS-6286R-A350
Anti-TMPRSS4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
TMPRSS4 is a member of the peptidase S1 family and contains 1 LDL receptor class A domain, 1 peptidase S1 domain and 1 SRCR domain. It is a probable membrane protease capable of activating ENaC and may process sodium channels in endothelial cells. TMPRSS4 is overexpressed in thyroid neoplasms, and splice variants in TMPRSS4 are thought to be linked with different cancers. Three named isoforms are produced by alternative splicing.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge