Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PRUNE Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6299R-CY5
BOSSBS-6299R-CY5EA 900 EUR InStock
BOSSBS-6299R-CY5
Anti-PRUNE Rabbit Polyclonal Antibody (Cy5®)
Antikörper
PRUNE, the human homologue of the Drosophila gene, is located in 1q21.3, a region highly amplified in human sarcomas, malignant tumours of mesenchymal origin. Human prune (h-prune), a phosphoesterase DHH family appertaining protein, physically interacts with nm23-H1, a metastasis suppressor gene. h-prune is involved in cellular motility and metastasis formation. Metastatic breast cancers were found to overexpress h-prune; h-prune was also found to be highly expressed in colorectal and pancreatic cancers. Hence h-prune is considered useful as a marker for tumor aggressiveness.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRUNE
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge