Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VGF Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6349R-CY3
BOSSBS-6349R-CY3EA 900 EUR InStock
BOSSBS-6349R-CY3
Anti-VGF Rabbit Polyclonal Antibody (Cy3®)
Antikörper
May be involved in the regulation of cell-cell interactions or in synatogenesis during the maturation of the nervous system. NERP peptides are involved in the control of body fluid homeostasis by regulating vasopressin release. Antimicrobial peptide VGF[554-577]: Has bactericidal activity against M. luteus, and antifungal activity against P. Pastoris.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Nerve growth factor inducible
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge