Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDK5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6411R-CY3
BOSSBS-6411R-CY3EA 902 EUR InStock
BOSSBS-6411R-CY3
Anti-CDK5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CDK5 (Cyclin Dependent Kinase 5) is serine/threonine kinase involved in synaptic regulation and neuronal development; phosphorylates synaptic protein Pctaire1; regulates acetylcholine receptor expression. CDK5 is a member of the cyclindependent kinase family of serine/threonine kinases. It is present in numerous mammalian tissues including kidney, testes, and ovary. Its activity is detected almost exclusively in brain extracts. Neuronal and muscle cells contain the highest amount of this protein. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity, but Cdk5 is not activated by cyclins. Instead, the p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 interacts with D1 and D3 type G1 cyclins and can phosphorylate histone H1, TAU, MAP2 and NF-H and NF-M. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDK5
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge