Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Lectin LCA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6439R-FITC
BOSSBS-6439R-FITCEA 900 EUR InStock
BOSSBS-6439R-FITC
Anti-Lectin LCA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Lens culinaris agglutinin is composed of four subunits - two of about 17 kDa and two of 8 kDa. LCA recognizes sequences containing α-linked mannose residues but recognizes additional sugars as part of the receptor structure, giving it a narrower specificity than Con A. An α-linked fucose residue attached to the N-acetylchitobiose portion of the core oligosaccharide markedly enhances affinity. LCA has been found to be one of the most effective agents in preventing skin allograft rejection in model systems. LCA has been used to purify numerous glycoproteins, including immunoglobulins, histocompatibility antigens, and α2-macroglobulin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Lectin LCA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge