Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C14ORF166 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6514R-CY3
BOSSBS-6514R-CY3EA 900 EUR InStock
BOSSBS-6514R-CY3
Anti-C14ORF166 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CGI-99 is a 244 amino acid protein that localizes to the nucleus, as well as to the cytoplasm, and belongs to the UPF0568 family. Expressed in a variety of tissues with particularly high levels in heart and skeletal muscle, CGI-99 functions as a homodimer that interacts with Ninein and is thought to regulate Ninein function. CGI-99 is overexpressed in brain tumor tissue, suggesting a role in tumor development and metastasis. The gene encoding CGI-99 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder ?-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C14ORF166
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge