Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-OSBP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6560R-CY5
BOSSBS-6560R-CY5EA 900 EUR InStock
BOSSBS-6560R-CY5
Anti-OSBP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Oxysterol-binding protein (OSBP) family of proteins consist of OSBP (OSBP1) and OSBP2 (ORP-4), which share a high overall similarity. OSBPs are involved in lipid metabolism and signal transduction, as well as vesicle transport, and can translocate to the periphery of Golgi membranes when they are bound to oxysterols. The OSBP protein transports sterols from lysosomes to the nucleus, where sterols downregulate the genes for HMG synthetase, HMG-CoA reductase and the low density lipoprotein receptor (LDLR). OSBP localizes to the cytosol and is widely expressed, while OSBP2 is mainly detected in testis, retina and fetal liver. The extracellular signal-regulated kinase (ERK) signaling pathway is controlled by OSBP via its cholesterol-binding properties. OSBP binds with a high affinity to 25-hydroxy-cholesterol (25-HC), a suppressor of cholesterol synthesis gene transcription in cultured cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OSBP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge