Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MRGPRX1/3/4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6606R-A680
BOSSBS-6606R-A680EA 893 EUR InStock
BOSSBS-6606R-A680
Anti-MRGPRX1/3/4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
There are four members (MRGX1-4) in the human MRGX familly. MRGX1 and MRGX2 receptors stimulate both G Alpha q- and G Alpha i -regulated pathways, while MRGX3 and MRGX4 receptors mainly activate G Alpha q-regulated pathways. G Alpha q proteins are involved in the calcium-signaling pathway downstream of the MRGX receptors. MRGX receptors are unique in that they are expressed in a subset of small dorsal root and trigeminal sensory neurons. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Potently activated by enkephalins including BAM22 (bovine adrenal medulla peptide 22) and BAM (8-22). BAM22 is the most potent compound and evoked a large and dose-dependent release of intracellular calcium in stably transfected cells. G(alpha)q proteins are involved in the calcium-signaling pathway.

Type: Primary
Antigen: MRGPRX1/3/4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/322
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge