Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6624R-A647
BOSSBS-6624R-A647EA 893 EUR InStock
BOSSBS-6624R-A647
Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge