Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCAF1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6735R-CY7
BOSSBS-6735R-CY7EA 1350 EUR InStock
BOSSBS-6735R-CY7
Anti-SCAF1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
SR-A1 (serine arginine-rich pre-mRNA splicing factor) is a member of the SR protein family, which interacts with the C-terminal domain of the large subunit of RNA polymerase II and involved in pre-mRNA splicing. The SR-A1 gene is located on chromosome 19q13.3-q13.4. Overexpression of SR-A1 is found in aggressive ovarian cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCAF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge