Anti-Cancer testis antigen 65 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-6746R-A680
BOSSBS-6746R-A680EA 893 EUR InStock
BOSSBS-6746R-A680
Anti-Cancer testis antigen 65 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
TULP2 is a member of a family of tubby-like (TULP) proteins which share a conserved C terminal region of approximately 200 amino acid residues. It is strongly expressed in testis and is expressed in the retina. It is also expressed in cancer cell lines.
Type: Primary
Antigen: Cancer testis antigen 65
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/520
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge