Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APIP Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6792R-CY5.5
BOSSBS-6792R-CY5.5EA 900 EUR InStock
BOSSBS-6792R-CY5.5
Anti-APIP Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The mammalian homologues of the key cell death gene CED 4 in C. elegans has been identified recently from human and mouse and designated Apaf1 (for apoptosis protease activating factor 1). Apaf1 binds to cytochrome c (Apaf2) and caspase 9 (Apaf3), which leads to caspase 9 activation. Activated caspase 9 in turn cleaves and activates caspase 3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. A new Apaf1 Interacting Protein (APIP) also known as CG129 and MMRP19, has been identified as a negative regulator of ischemic injury. APIP competes with Caspase 9 binding site of Apaf1. APIP is predicted to code for a 204 amino acid. An isoform of APIP, APIP2 encodes a 242 amino acid protein, which is an alternative splicing variant differing in its N terminus from APIP. APIP transcript is ubiquitously expressed in most adult tissue with high expression in skeletal muscle, heart, and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APIP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge