Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAPN12 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6798R-A647
BOSSBS-6798R-A647EA 893 EUR InStock
BOSSBS-6798R-A647
Anti-CAPN12 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Calpains are a family of cytosolic calcium activated cysteine proteases involved in a variety of cellular processes including apoptosis, cell division, modulation of integrin and cytoskeletal interactions, and synaptic plasticity. Calpain 12 was first described in the mouse, most strongly in the skin, and maps to mouse chromosome 7. Isoforms differ in the carboxyterminal ends, ending with aberrant domain III and lacking domain IV. Domains in the large subunit include the amino terminal domain I, the proteinase domain II, domain III, and the EF hand domain IV, making Calpain 12 most similar to calpains 1 and 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPN12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge