Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-beta COP Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6857R-A750
BOSSBS-6857R-A750EA 893 EUR InStock
BOSSBS-6857R-A750
Anti-beta COP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The Golgi complex is a key organelle where processing and sorting of newly synthesised proteins occurs. Membrane traffic from the endoplasmic reticulum (ER) to the Golgi complex and from the Golgi complex to the different final cellular destinations is believed to be mediated by carrier vesicles. Two populations of coated vesicles mediate biosynthetic membrane traffic between the different membrane-bound compartments. Clathrin-coated vesicles carry proteins to endocytic organelles and secretory granules, whilst non-clathrin-coated vesicles are involved in intra-Golgi transport and transport from the ER to the Golgi complex. Beta COP is a member of a set of protein which are believed to associate with the non-clathrin coated vesicles. Golgi-derived non-clathrin-coated vesicles are believed to act as bulk carriers, whereas clathrin-coated vesicles carry a selective cargo of membrane proteins.

Type: Primary
Antigen: beta COP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 531-630/953
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge