Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KDM2A Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6943R
BOSSBS-6943REA 725 EUR InStock
BOSSBS-6943R
Anti-KDM2A Rabbit Polyclonal Antibody
Antikörper
F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and protein recruitment. F-box proteins are members of a large family that regulate the cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, for degradation by the proteasome after ubiquitination. FBL11, also known as FBXL11 (F-box and leucine-rich repeat protein 11), CXXC8, KDM2A, JHDM1A (JmjC domain-containing histone demethylation protein 1A) or LILINA, is a 1,162 amino acid member of the F-box protein family that contains one F-box domain and localizes to the nucleus. Expressed ubiquitously with highest expression in testis, ovary and brain, FBL11 functions to demethylate the Lys-36 residue of histone H3, thereby modulating the histone code. Additionally, FBL11 is thought to promote the ubiquitination and subsequent degradation of various phosphorylated proteins. Three isoforms of FBL11 exist due to alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge