Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RUBISCO Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6988R-CY3
BOSSBS-6988R-CY3EA 897 EUR InStock
BOSSBS-6988R-CY3
Anti-RUBISCO Rabbit Polyclonal Antibody (Cy3®)
Antikörper
RuBisCO catalyzes two reactions: the carboxylation of D-ribulose 1,5-bisphosphate, the primary event in carbon dioxide fixation, as well as the oxidative fragmentation of the pentose substrate in the photorespiration process. Both reactions occur simultaneously and in competition at the same active site.Sequence similarities: Belongs to the RuBisCO large chain family. Type I subfamily.Post-translational modifications: The disulfide bond which can form between Cys-247 in the large chain dimeric partners within the hexadecamer appears to be associated with oxidative stress and protein turnover (By similarity). The disulfide bonds reported in 1RBO may be the result of oxidation during crystallization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RUBISCO
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge