Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CSTF2T Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-6995R-A555
BOSSBS-6995R-A555EA 893 EUR InStock
BOSSBS-6995R-A555
Anti-CSTF2T Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. CstF-64 contains an RNA binding domain and is responsible for the RNA binding activity of CstF. CstF-64 is expressed in all somatic cells and in pre- and postmeiotic, but not meiotic, germ cells. However, a large variant of CstF-64, called t CstF-64, is abundantly expressed in meiotic and postmeiotic cells in the testis and to a lesser extent in the brain, and promotes the germ cell pattern of polyadenylation. The gene encoding CstF-64 (designated CSTF2) maps to the X chromosome, whereas t CstF-64 is encoded by an autosomal gene. The increase in CstF-64 concentration during B cell activation switches IgM heavy chain mRNA expression from membrane-bound to secreted forms, suggesting that CstF-64 plays a key role in regulating IgM heavy chain expression during B cell differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSTF2T
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge