Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IFN-Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7021R-A750
BOSSBS-7021R-A750EA 893 EUR InStock
BOSSBS-7021R-A750
Anti-IFN-Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Interferon-_ (IFN-_, also known as IFN-alpha, IFN-a, IFNA and Interferon-alpha) is a type I interferon and exists as 13 subtypes: IFN-_ 1, 2, 4, 5, 6, 7, 8, 10, 13, 14, 16, 17 and 21. They are mainly produced by virus-infected cells after sensing the presence of danger signals such as viral DNA or RNA. The binding of IFN-_ to its receptor initiates an anti-viral response, but also anti-tumor effects have been shown. A key producer of type I interferons is the plasmacytoid dendritic cell.

Type: Primary
Antigen: IFN-Alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-189/189
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge