Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MEOX2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7031R-HRP
BOSSBS-7031R-HRPEA 1350 EUR InStock
BOSSBS-7031R-HRP
Anti-MEOX2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Closely related homeobox proteins, MOX-1 and MOX-2, belong to a family of nonclustered, diverged homeobox genes that are expressed in overlapping patterns in the paraxial mesoderm and its derivatives. MOX-1 and MOX-2 function transiently in the formation of mesodermal and mesenchymal derivatives. MOX-1 and MOX-2 are implicated in the early steps of mesoderm formation during gastrulation. In addition, the MOX proteins are also involved in somatic differentiation. Significantly, MOX-1 associates more strongly with Pax-1, whereas MOX-2 preferentially associates with Pax-3. Specifically, expression of MOX-2 (also known as mesenchyme homeobox 2 or GAX), has been shown to be critical in axial skeleton development. MOX-2 is not needed for the migration of myogenic precursors into the limb bud, but it is essential for normal appendicular muscle formation and for the normal regulation of myogenic genes. MOX-2 is expressed in placental tissue. The human MEOX2 gene maps to chromosome 7p21.1 and encodes the MOX-2 protein. Mutations in the gene may be involved in craniofacial and/or skeletal abnormalities.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: MEOX2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge