Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FAIM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7061R-HRP
BOSSBS-7061R-HRPEA 900 EUR InStock
BOSSBS-7061R-HRP
Anti-FAIM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
FAIM1 (or Fas Apoptotic Inhibitory Molecule 1) is a recently discovered negative regulator of apoptosis. FAIM1 has no significant regions of homology to other gene products that modulate Fas killing. It plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells. Overexpression of FAIM1 diminishes sensitivity to Fas mediated apoptosis of B and non B cell lines. FAIM1 is highly evolutionarily conserved and is widely expressed in murine tissues, suggesting that FAIM1 plays an important role in cellular physiology.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FAIM1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge