Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Nucleolar protein 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7080R-A680
BOSSBS-7080R-A680EA 893 EUR InStock
BOSSBS-7080R-A680
Anti-Nucleolar protein 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD)bcontaining molecules and a caspase family of proteases. CARD containing cell death regulators include RAIDD, RICK, BCL10, Apaf 1, caspase 9 and caspase 2. Apoptosis repressor with CARD is a CARD domain containing protein that interacts with caspase 2 and 8 to inhibit enzymatic activity of caspase 8. Apoptosis repressor with CARD suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR 1, and DR3. The mRNA of Apoptosis repressor with CARD is primarily expressed in skeletal muscle and cardiac tissue. The nuclear isoform (1/Nop30) may be involved in RNA splicing and the cytoplasmic isoform (2/Myp) may inhibit apoptosis.

Type: Primary
Antigen: Nucleolar protein 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/220
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge