Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TP53I11 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7091R-CY7
BOSSBS-7091R-CY7EA 900 EUR InStock
BOSSBS-7091R-CY7
Anti-TP53I11 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
TP53I11 (Tumor protein p53 inducible protein 11), also referred to as PIG11 (p53-induced gene 11), is an early transcriptional target of tumor suppressor p53. It is up-regulated in the induction of apoptosis or cell growth inhibition by multiple chemopreventive agents. However, its biological role remains unclear.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53I11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge