Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PLSCR3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7099R-FITC
BOSSBS-7099R-FITCEA 900 EUR InStock
BOSSBS-7099R-FITC
Anti-PLSCR3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
PLSCR3 (phospholipid scramblase 3) may mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. Members of this family, PLS1 and PLS3 are both substrates of Protein kinase C (PKC) delta and are phosphorylated by PKC-delta during apoptosis. PKC-delta translocates to mitochondria during apoptosis and phosphorylates PLS3. Overexpression of PLS3 in the HEK293 cells enhanced apoptosis induced by UV-irradiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLSCR3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge