Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RAD23 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7121R-FITC
BOSSBS-7121R-FITCEA 900 EUR InStock
BOSSBS-7121R-FITC
Anti-RAD23 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The RAD23 gene of Saccharomyces cerevisiae is required for excision-repair of UV damaged DNA (1). RAD23 resembles the other DNA repair genes, RAD2, RAD6, RAD7, RAD18, and RAD54, all of which also exhibit increased transcription in response to DNA damage and during meiosis (2). RAD23 encodes a nuclear protein containing a ubiquitin-like domain required for biological function (3). RAD23 bears a ubiquitin-like domain at its amino terminus and this ubiquitin-like domain affects protein function in a nonproteolytic manner (3). Rad23 is a highly conserved protein involved in nucleotide excision repair (NER) that associates with the proteasome via its amino-terminus (4). Its carboxy-terminal ubiquitin-associated domain is evolutionarily conserved from yeast to humans (4). In addition to a role in DNA repair events in yeast, several lines of evidence indicate that the Rad23 protein may regulate the activity of the 26 S proteasome (5).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAD23
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge