Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FKBP6 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7126R
BOSSBS-7126REA 1090 EUR InStock
BOSSBS-7126R
Anti-FKBP6 Rabbit Polyclonal Antibody
Antikörper
FKBP6 is a 327 amino acid protein that, like other PPIases, accelerate the folding of proteins. As a component in synaptonemal complexes, FKBP6 is involved in homologous chromosomes pairing and male infertility in mice. There has been some suggestion that FKBP6 may play a role in modifying the susceptibility to idiopathic spermatogenic impairment in humans. Ubiquitously expressed in all tissues, FKBP6 is present at highest levels in testis, liver, kidney, skeletal muscle and heart. The gene encoding FKBP6 maps within a region of human chromosome 7 that has been implicated in Williams-Beuren syndrome, a rare developmental disorder involving abnormalities of the cardiovascular and musculoskeletal systems. Hemizygous deletion of FKBP6 may contribute to hypercalcemia and growth delay in Williams-Beuren syndrome.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FKBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge