Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLC25A28 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7157R-CY5.5
BOSSBS-7157R-CY5.5EA 900 EUR InStock
BOSSBS-7157R-CY5.5
Anti-SLC25A28 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Mitoferrin 2, also known as MRS3/4 (mitochondrial RNA-splicing protein 3/4 homolog), mitochondrial iron transporter 2, NPD016, MRS4L or SLC25A28 (solute carrier family 25 member 28), is a 364 amino acid multi-pass membrane protein of the mitochondrial inner membrane that mediates iron uptake. Mitoferrin 2 is thought to play a role in heme synthesis of hemoproteins and iron-sulfur cluster assembly. Ubiquitously expressed, Mitoferrin 2 is found at high levels in skeletal muscle, heart, placenta, kidney, lung, liver, brain and pancreas. Mitoferrin 2 is a member of the mitochondrial carrier family and undergoes alternative splicing events to produce four isoforms. Mitoferrin 2 contains three solcar repeats and is encoded by a gene that maps to human chromosome 10q24.2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A28
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge