Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Pantophysin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7411R-HRP
BOSSBS-7411R-HRPEA 900 EUR InStock
BOSSBS-7411R-HRP
Anti-Pantophysin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The MARVEL domain is a 130 amino acid motif that contains four transmembrane helices, both of which have cytoplasmic N- and C-terminal regions. MARVEL domain-containing proteins are thought to participate in tight junction regulation, the biogenesis of vesicular transport carriers and in cholesterol-rich membrane apposition events. Pantophysin, also known as SYPL1 (synaptophysin-like protein 1) or H-SP1, is a 259 amino acid multi-pass membrane protein that localizes to melanosomes and vesicles, as well as to the cytoplasm, and contains one MARVEL domain. Expressed as multiple alternatively spliced isoforms, pantophysin is present in tissues throughout the body where it may play a role in vesicle trafficking and protein transport. The gene encoding pantophysin maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Pantophysin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge