Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-POLD2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7416R-CY5.5
BOSSBS-7416R-CY5.5EA 900 EUR InStock
BOSSBS-7416R-CY5.5
Anti-POLD2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases (1). In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches (2). These exonucleases include the family of DNA polymerases (3). DNA pol α, β, ∂, and e are involved in DNA replication and repair (4). DNA pol ∂ and DNA pol e are multisubunit enzymes, with DNA pol ∂ consisting of two subunits p125, which interacts with the sliding DNA clamp protein PCNA, and p50 (5). The nuclear-encoded DNA pol © is the only DNA polymerase required for the replication of the mitochondrial DNA (6). DNA pol Ω is ubiquitously expressed in various tissues and mediates the cellular mechanism of damage-induced mutagenesis (7). DNA pol œ is a DNA polymerase-helicase that binds ATP and is involved in the repair of interstrand crosslinks (8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POLD2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge