Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SAP155 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7417R-A750
BOSSBS-7417R-A750EA 893 EUR InStock
BOSSBS-7417R-A750
Anti-SAP155 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
SAP 155 is a 1,304 amino acid member of the SF3B1 family and contains eleven HEAT repeats. Localised to nuclear speckles and also to the cytoplasm during mitosis, SAP 155 is a subunit of the SF3B splicing factor. The SF3B splicing factor is a U2 snRNP-associated protein complex essential for spliceosome assembly. SF3B contains the spliceosomal proteins SAP 49, SAP 130, SAP 145 and SAP 155. Concomitant with splicing catalysis, SAP 155 is phosphorylated at its N-terminal Thr-Pro dipeptide motifs by Dyrk1A and cyclin E/Cdk2. This modification of SAP 155 is vital for a functional spliceosome as it is an essential event in the basic splicing reaction. Due to alternative splicing events, various SAP 155 isoforms are produced.

Type: Primary
Antigen: SAP155
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/1304
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge