Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BATF Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7463R-A647
BOSSBS-7463R-A647EA 893 EUR InStock
BOSSBS-7463R-A647
Anti-BATF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
B-ATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of B-ATF mediates dimerization with members of the Jun family of proteins. The B-ATF protein does not homodimerize efficiently, but rather forms a heterodimer preferentially with c-Jun. The B-ATF/c-Jun protein complex can interact with DNA containing a consensus binding site for AP-1, suggesting that B-ATF functions as a tissue-specific modulator of the AP-1 transcription complex in human cells. B-ATF also associates with IFP35, a leucine zipper protein that translocates to the nucleus following IFN treatment. The gene encoding B-ATF, also designated SFA-2, is strongly expressed in mature T and B lymphocytes, and is up-regulated after transformation by human T-cell leukemia virus type I.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BATF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge