Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADGRB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7513R-FITC
BOSSBS-7513R-FITCEA 900 EUR InStock
BOSSBS-7513R-FITC
Anti-ADGRB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
BAI 1 protein is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain. Reduced or no expression is observed in some glioblastoma cell lines and cancer tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge