Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-INSRR Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7531R-A488
BOSSBS-7531R-A488EA 893 EUR InStock
BOSSBS-7531R-A488
Anti-INSRR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The insulin receptor related receptor (IRR) is a heterotetrameric transmembrane receptor composed of two alpha and 2 beta chains linked by disulfide bonds. The alpha chains contribute to the formation of the ligand-binding domain, while the beta chains carry the kinase domain. Member of the insulin RTK family, IRR shares high homology with the insulin (IR) and the insulin-like growth factor-1 receptor (IGF-1R), but doesn’t bind any of IR and IGF-1R known ligands. In contrast to the widespread patterns of expression to IR and IGF-1R, IRR demonstrates a very restricted cellular distribution in a subset of tissues of neuronal origin and its biological functions are still unknown.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: INSRR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge