Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CPB2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7547R-A488
BOSSBS-7547R-A488EA 893 EUR InStock
BOSSBS-7547R-A488
Anti-CPB2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This protein belongs to a family of Zn-containing metallocarboxypeptidases specific to C-terminal lysine and arginine residues. It circulates in plasma as a zymogen with molecular weight of 55 kDa (401 amino acid residues; pI 5.0). Being activated by thrombin-thrombomodulin complex during blood coagulation, it exerts carboxypeptidase activity. Activated carboxypeptidase B2 removes C-terminal lysine residues from fibrin, which is necessary for plasminogen binding to fibrin. This prevents plasminogen from activation into plasmin and retards the lysis of a fibrin clot. The concentration in plasma of healthy people is 5-10 ug/ml. High plasma levels were found in patients with stable angina pectoris and angiographically verified coronary artery disease. Elevated concentration in blood is considered as a risk factor for venous thrombosis. A deficiency might contribute to the severity of bleeding disorders in hemophilias A and B, and decreased levels are found in chronic liver disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge